NHS Ester, SureLight® 488 molecular biology stain
Supplier: VWR International
76311-696 76311-698
76311-696EA 71.31 CAD
76311-696 76311-698
NHS Ester, SureLight® 488 molecular biology stain
NHS Ester
SureLight® 488 NHS Ester is a bright green fluorescent dye with excitation suited to the 488nm laser line, fluorescent microscopy and flow cytometry. It offers a much more stable signal generation in imaging and flow cytometry than FITC and is half the cost of Alexa Fluor® 488 with no royalties or licensing fees.
  • Excitation max 486 nm
  • Emission max 517 nm
The NHS-ester version is an efficient amine reactive probe that can be used to label proteins, antibodies, haptens and other primary amine containing molecules.