Alumina SPE Columns, Honeywell Burdick & Jackson
Supplier: Burdick & Jackson
9114 9124 9128 9130 9134 9138
CABJ9114PK 625.62 CAD
CABJ9114 CABJ9124 CABJ9128 CABJ9130 CABJ9134 CABJ9138
Alumina SPE Columns, Honeywell Burdick & Jackson
Chromatography Columns
These SPE columns are made from medical-grade polypropylene with PE frits.
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar.