LB Agar Plates with 100 µg/ml Kanamycin, Teknova
Supplier: TEKNOVA CA
L1027
L5027
L3027
CA100216-592PK
322.54
CAD
CA100216-592
CA100217-258
CA100218-234
LB Agar Plates with 100 µg/ml Kanamycin, Teknova
Media for Microbiology
Prepared Media for Microbiology
Ready to use LB agar plates (Miller formulation) for the growth of recombinant E. coli. All plates are made with the highest quality products and tested with four strains of E. coli.
Ingredients: 1.0% tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 100 µg/ml kanamycin.