LOCTITE® 680 Retaining Compounds, Henkel Corporation
Supplier: Henkel
Loctite®
1835205
76313-912EA 409.98 CAD
76313-912
LOCTITE® 680 Retaining Compounds, Henkel Corporation
Adhesives
Designed for the bonding of cylindrical fitting parts, particularly where low viscosity is required. This compound prevents loosening and leakage from shock and vibration.
  • Green in color
  • High strength
The product prevents loosening and leakage from shock and vibration. It provides robust curing performance and works on active metals (mild steel) and on passive substrates (stainless steel and plated surfaces).
Ordering information: Ten pieces per case. Must be ordered in case quantity.
Packaging: 10 ml bottle, 0.081 kg Package Weight, 2.8L×2.8W×8.9H cm Packaging Dimension.