Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
33665-10L-VP
33665-6X1L
33665-5L-VP
33666-10L-VP
33666-6X1L
33666-6X500ML
33666-5L-VP
33667-6X1L
33668-10L-VP
33668-6X1L
33665-6X500ML
BJ33665-10L-VPEA
573.62
CAD
BJ33665-10L-VP
BJ33665-1L
BJ33665-500ML
BJ33665-5L-VP
BJ33666-10L-VP
BJ33666-1L
BJ33666-500ML
BJ33666-5L-VP
BJ33667-1L
BJ33667-500ML
BJ33667-5L-VP
BJ33668-10L-VP
BJ33668-1L
BJ33668-500ML
BJ33665-6X500ML
Buffer Solution, Color-Coded, Fluka™, Honeywell Research Chemicals
Buffers
pH Calibration Buffers
These buffer solutions are color-coded for easy identification and the pH 4 and 7 version have fungicide additive.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...