Aluminum Laboratory Jacks
Supplier: GSC INTERNATIONAL SE
LBJK150 LBJK200
470313-328EA 130.7 CAD
470313-328 470313-330
Aluminum Laboratory Jacks
Laboratory Jacks
These aluminum and stainless-steel laboratory jacks provide a helpful platform for use in the lab.
Two sizes to choose from:
150 mm (~5.9") by 150 mm Platform, Maximum Height of 260 mm (~10.25"), Holds up to 25 kg (~55lbs.)
200 mm (~7.9") by 200 mm Platform, Maximum Height of 280 mm (~11"), Holds up to 30 kg (~66lbs.)