Water Standards, CRM, Karl Fischer Titration, Hydranal™, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
34424-10G 34425-80ML 34426-40ML
BJ34424-10GEA 205.1 CAD
BJ34424-10G BJ34425-80ML BJ34426-40ML
Water Standards, CRM, Karl Fischer Titration, Hydranal™, Fluka™, Honeywell Research Chemicals
Standards Water Determination (by Karl Fischer method) Standards
Certified reference material for Karl Fischer titration, exact value on certificate of analysis.
  • For volumetric and coulometric Karl Fischer applications
  • Manufactured according to ISO 17034 requirements
  • Certificate of Analysis showing the exact water content, expanded uncertainty plus comprehensive product information and instruction for use
  • Lot-related stability and homogeneity studies
  • Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Convenient packaging
Hydranal CRMs serve as a reference point. They are traceable to high-purity materials of well-established purity and/or SI units. Additionally they are verified against internationally accepted standards from national metrological institutes such as NIST (National Institute of Standards & Technology, USA) or NMIJ (National Metrology Institute of Japan). In addition to traceability to accepted references, Hydranal CRMs have a well-defined mass fraction and highly accurate measurement of uncertainty which can be found in the Certificate of Analysis (CoA).
Certifications: Hydranal CRMs are tested under ISO/IEC 17025 and comply with ISO 17034 requirements.