Acetate Buffer Solution, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
31048-1L 31048-6X500ML 31103-6X1L
BJ31048-1LEA 118.36 CAD
BJ31048-1L BJ31048-500ML BJ31103-1L BJ31103-6X1L
Acetate Buffer Solution, Fluka™, Honeywell Research Chemicals
Buffers pH Calibration Buffers
This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable