Ultraviolet Fluorometer Cuvettes, FireFlySci
Supplier: FIREFLYSCI, INC.
11FLUV5 11FLUV10 11FLUV20 11FLUV40
76015-378EA 1055.5 CAD
76015-378 76015-380 76015-382 76015-384
Ultraviolet Fluorometer Cuvettes, FireFlySci
Cuvettes Spectrophotometer Cuvettes
Ultraviolet Fluorometer Cuvettes have four windows with base polished and quartz cap.