Helix® 1071 Series Gloves, HexArmor
Supplier: HexArmor
1071-L(9) 1071-M(8) 1071-S(7) 1071-XL(10) 1071-XS(6) 1071-XXL(11) 1071-XXS(5) 1071-XXXL(12)
76318-056CS 783.55 CAD
76318-056 76318-058 76318-060 76318-062 76318-064 76318-066 76318-068 76318-070
Helix® 1071 Series Gloves, HexArmor
Gloves
Helix® 1071 series gloves are made with a 15-gauge nylon and fiberglass blended shell. They feature a smooth microfoam nitrile coating for excellent comfort and dexterity, allowing hands to move freely and safely on the job.
  • High-performance nylon and fiberglass blend shell
  • Microfoam nitrile palm coating provides superior grip and abrasion resistance
  • Seamless construction for enhanced comfort and breathability
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
Helix® series gloves offer a wide variety of gripMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Powdered/Powder-free Powder-free
Liner material Nylon/fibreglass
Finish Microfoam Nitrile Palm
Cuff style Knit wrist