Reagent Reservoirs, Thermo Scientific
Supplier: Thermo Scientific
8094 8085 8086 8093-11 8095 8096-11
76310-566CS 277.71 CAD
76310-566 76310-568 76310-570 76310-572 76310-574 76310-576
Reagent Reservoirs, Thermo Scientific
Reagent Reservoirs
Features trough-within-a-trough maximizes amount of liquid accessible to pipette tips when using small amounts of reagent.
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Maximizes recovery if using only single channel and small amount of reagent
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
Divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other.
Packaging: Sterile reservoirs packaged in snap-and-tear bags maintains sterility.