T.S.C. Supplement (Clostridium perfringens T.S.C. Supplement), HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
FD014-5VL FD014-4X5VL
CA89128-830EA 463.56 CAD
CA89128-830 CA71005-592
T.S.C. Supplement (Clostridium perfringens T.S.C. Supplement), HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement, recommended for the selective isolation of Clostridium perfringens.