Modified Duncan Strong (DS) Medium, Vegetable Based, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV1237-500G MV1237-2.5KG MV1237-5KG
CA61000-580EA 2551.42 CAD
CA61000-580 CA89227-894 CA89227-896
Modified Duncan Strong (DS) Medium, Vegetable Based, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Modified Duncan Strong (DS) medium is used for isolation and differentiation of Clostridium perfringens from other clostridia from foods on the basis of raffinose fermentation.
  • Formulated as per Duncan and Strong
  • Isolation and differentiation of C. perfringens from other Clostridia from foods on the basis of raffinose fermentation
  • Rapid detection of the Clostridium perfringens enterotoxin
Clostridium perfringens, a gram-positive, rod shaped, anaerobic, spore-forming bacteria, is the major cause of food poisoning in humans.
Modified Duncan Strong (DS) Medium is formulated as per Duncan and Strong, and is recommended by APHA for the isolation and differentiation of C. perfringens from other Clostridia from foods on the basis of raffinose fermentation. It is also recommended for the rapid detection of the Clostridium perfringens enterotoxin.
Proteose peptone and yeast extract provide nitrogenous compounds and other nutrients for bacterial growth. Sodium thioglycollate helps to create anaerobic conditions suitable for clostridial growth. Disodium phosphate acts as a buffering agent. Raffinose in the medium is fermented by C. perfringens to produce acid within 72 hours, but not by culturally similar species like Clostridium baratii, Clostridium celatum, and more.