Capping Reagent kit contains 40% Acetic Anhydride/ 60% Acetonitrile (BR643) and 60% 2,6-Lutidine / 40% Acetonitrile (BR645), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
BR648-1
BR648-2.5
CABJBR648-1KT
771.86
CAD
CABJBR648-1
CABJBR648-2.5
Capping Reagent kit contains 40% Acetic Anhydride/ 60% Acetonitrile (BR643) and 60% 2,6-Lutidine / 40% Acetonitrile (BR645), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Reagents are specifically processed and purified to ensure the high synthesis efficiency
Contains 40% Acetic Anhydride/ 60% BioSyn™, for DNA/RNA Synthesis, Acetonitrile (BR643) and 60% 2,6-Lutidine / 40%Acetonitrile (BR645)
- Ideal for oligonucleotide synthesis
- Extremely low water content
- High purity reagents
- Low amine specifications
Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.