5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 5-(Ethylthio)tetrazole5-Ethyltio-1H-Tetrazole

BR725-2.5GL BR725-20 BR725-1
BJBR725-2.5GLCS 26534.43 CAD
BJBR725-2.5GL BJBR725-1 BJBR725-20
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazole (ETT)

Reagents are specifically processed and purified to ensure the high synthesis efficiency


  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications


Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.

Formula: C₃H₆N₄S
MW: 130.17 g/mol
Boiling Pt: 82 °C (main component)
Melting Pt: –46 °C (main component)
Density: 0.809 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00068733
CAS Number: 89797-68-2
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR