Starch, soluble A.R.

Supplier: HIMEDIA LABORATORIES PVT. CA

Synonyms: StarchVitex® iodine indicatorAmylodextrin

RM3029-500G RM3029-500G
CA95026-656EA 514.42 CAD
CA95026-656 95026-656
Starch, soluble A.R.
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance White Powder
pH (2% in water at 25 °C) 6.0 - 7.5
Clarity 1% in Boiling Water gives Clear Solution
FTIR (KBr disc) Matches with the Standard Pattern
Chloride (Cl) ≤0.005%
Loss on Drying ≤15.0%
Sulphated Ash ≤0.4%
Reducing Substances ≤0.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR