Starch Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M107-500G M107-2.5KG M107-5KG
CA95020-426EA 988.7 CAD
CA95020-426 CA89222-824 CA89222-838
Starch Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Starch agar is used for the detection of starch hydrolyzing microorganisms.