Sample Spoons, Bürkle
Supplier: BURKLE INC
5320-0210 5320-0300 5320-0400 5320-0500 5320-0150 5320-0180
76319-782EA 24.73 CAD
76319-782 76319-784 76319-786 76319-788 76319-854 76319-856
Sample Spoons, Bürkle
Spoons
Sampling Spoons are designed for withdrawing small quantities of samples.
  • Made of AISI 304 (1.4301) stainless steel
  • Largely chemical resistant (not against acids and alkalis)
This specially shaped spoon allows users to reach almost every corner in a container, while preventing samples from trickling out.
Order Now
SPECIFICATIONS
Environmentally Preferable