HiVeg™ Streptococcus thermophilus Isolation Agar, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV948-500G MV948-2.5KG MV948-5KG
CA61001-482EA 1320.26 CAD
CA61001-482 CA89229-458 CA89229-460
HiVeg™ Streptococcus thermophilus Isolation Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for determining the ratio of Streptococcus thermophilus and Lactobacillus bulgaricus in yoghurt.
  • Reconstitution: 42.0 g/l
  • Sterilization: 121 °C/15 minutes
Caution: Storage conditions: dry medium - below 30 °C, prepared medium 2 to 8 °C.