Indo-1 AM ≥96% (by HPLC) fluorescent indicator, for calcium determination

Supplier: Biotium

Synonyms: 1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylester, Indo-1-(acetoxymethyl) ester, 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, Indo-1 AM Ester

50043 50043-1 50044
89139-228EA 406.57 CAD
89139-228 89139-230 89139-232
Indo-1 AM ≥96% (by HPLC) fluorescent indicator, for calcium determination
Indo-1 AM

Indo-1, AM ester is membrane-permeant and thus can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate cell loading. Indo-1-AM Ester itself does not bind Ca2+ , but it is readily hydrolyzed to indo-1 by endogenous esterases once it enters cells.


  • Absorption/Emission 356 nm/478 nm (no Ca2+); 330 nm/400 nm (high Ca2+)
  • Extinction Coefficient 33,000 M -1 cm -1 (no Ca2+); 33,000 M -1 cm -1 (high Ca2+)
  • Cell permeability: Membrane permeant
  • Light yellow solid soluble in DMSO
  • Store desiccated at -20°C and protect from light, especially in solution

Formula: C₄₇H₅₁N₃O₂₂
MW: 1009.91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036955
CAS Number: 112926-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR