Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination
Supplier: Biotium
50037
50038
50039
89139-216EA
537.15
CAD
89139-216
89139-218
89139-220
Furaptra (Mag-Fura-2) AM Ester ≥95% (by TLC) fluorescent indicator, for calcium determination
Furaptra (Mag-Fura-2) AM Ester
Furaptra, AM ester is a membrane-permeant form of Furaptra (Mag-Fura-2). The indicator is loaded into cells via simple incubation
- Absorption/Emission 366nm/475nm (EtOAc)
- Extinction Coefficient 31,000
- Cell permeability: Membrane permeant
- Colors: Green
- Light yellow solid soluble in DMSO
- Store at -20°C and protect from light
Formula:
C₃₀H₃₀N₂O₁₉ Storage Temperature: Freezer |
CAS Number:
130100-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...