WHEATON® Battery Powered Crimpers and Decappers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W225808 W225813 W225814 W225815 W225811 W225812 W225820 W225821 W225822
76303-042CS 2081.05 CAD
76303-042 76303-044 76303-046 76303-048 76303-050 76303-052 76303-054 76303-056 76303-058
WHEATON® Battery Powered Crimpers and Decappers, DWK Life Sciences
Crimpers, Cappers and Decappers
These battery powered crimpers and decappers fit standard laboratory vials and seals. They come with a long-lasting battery with lithium ion technology, which allows for up to 800 crimps per charge.
  • Powerful motor with a fast cycle
  • Cushioned ergonomic handle reduces hand fatigue
  • Polished crimping jaws provide consistent sealing
  • Can be operated while charging
A warning light indicates when recharging is necessary. The tool can be recharged in one to two hours with the universal power supply included.
The electronically adjustable cycle is accurate and reproducible and can be adjusted with the control pad on top of the unit. The tools are ergonomically designed to reduce strain and arm injury associated with many manual crimping tools.