Neomycin trisulfate salt hydrate, ≥ 600 mg/mg (dry basis) USP

Danger

00224-100G 00224-25G
CH00224-100GEA 71.1 CAD
CH00224-100G CH00224-25G
Neomycin trisulfate salt hydrate, ≥ 600 mg/mg (dry basis) USP
Neomycin trisulfate salt hydrate
Formula: C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150829
CAS Number: 1405-10-3

Order Now

Specification Test Results

Appearance A White of Yellowish-White Powder
pH 5.0 - 7.5
Loss on Drying ≤8%
Assay ≥600 mcg/mg (Dried basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR