Permanent Deep Freeze Labels for Thermal Transfer Printers
Supplier: GA INTERNATIONAL INC
807656049007 807656049014 807656051468 807656051482 807656051802 807656051819 807656050720 807656050782 807656046457 807656046464 807656046785 807656046815 807656046907 807656046914 807656047461 807656047478 807656047546 807656047607 807656047706 807656047713 807656047744 807656047751 807656047928 807656047942 807656048543 807656048550 807656048567 807656048673 807656048802 807656048819 807656049045 807656049076 807656050188 807656050195 807656050386 807656050393 807656050485 807656050546 807656051864 807656051932 807656050881 807656050898 807656046945 807656047003 807656051048 807656047508 807656047515 807656051147 807656051154 807656051208 807656051246 807656047904 807656047911 807656048000 807656048017 807656048345 807656048352 807656050140 807656050164 807656051321 807656051383 807656051055 807656051543 807656051550 807656048369 807656048376 807656049106 807656049113 807656049885 807656049977
76316-064RL 283.99 CAD
76316-064 76316-066 76316-068 76316-070 76316-072 76316-074 76316-076 76316-078 76316-080 76316-082 76316-084 76316-086 76316-088 76316-090 76316-092 76316-094 76316-096 76316-098 76316-100 76316-102 76316-104 76316-106 76316-108 76316-110 76316-112 76316-114 76316-116 76316-118 76316-120 76316-122 76316-124 76316-126 76316-128 76316-130 76316-132 76316-134 76316-136 76316-138 76316-140 76316-142 76316-144 76316-146 76316-148 76316-150 76316-152 76316-154 76316-156 76316-158 76316-160 76316-162 76316-164 76316-166 76316-168 76316-170 76316-172 76316-174 76316-176 76316-178 76316-180 76316-182 76316-184 76316-186 76316-208 76316-210 76316-212 76316-214 76316-216 76316-218 76316-220 76316-222
Permanent Deep Freeze Labels for Thermal Transfer Printers
Labels
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
  • Temperature range: –80 to +110 °C (–112 to +230 °F)
  • Ideal for long-term storage in deep-freeze conditions (–80 °C)
  • No cure time; can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal transfer printer brands
  • Available in a large range of sizes,
More Product Information