ProSolo Digital Water Quality Meter Kits, ODO/T, YSI
Supplier: YSI
626663 626664
76308-058EA 3526.12 CAD
76308-058 76308-060
ProSolo Digital Water Quality Meter Kits, ODO/T, YSI
Electrochemistry Meters Oxygen Meters
These kits include a ProSolo dissolved oxygen meter, an ODO/T probe assembly with cable, and a soft-sided carrying case.
  • Optical dissolved oxygen sensor technology
  • Military spec (MS) connector and weighted, stainless steel probe guard
  • Color display and backlit keypad
  • Long-life rechargeable lithium-ion battery
  • 100 user-defined sites and data ID tags
  • Onboard memory with capacity for >100,000 data sets
  • Data export directly to USB flash drive
  • 400 detailed calibration records can be stored and are available to view and download
Certifications: CEC, CE; RoHS; IP-67; WEEE; FCC; UN Part III, Section 38.3, Test methods for lithium-ion batteries (Class 9).
Ordering information: Includes Digital ProSolo Handheld, cable, sensor and carrying case. Three-year warranty on handheld, two years on cable, probe, and ODO sensor cap.