ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
Supplier: YSI
627150-1 627150-4 627150-10 627150-20 627150-30 627150-50 627150-100
76308-044EA 2757.64 CAD
76308-044 76308-046 76308-048 76308-050 76308-052 76308-054 76308-056
ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
Electrodes Dissolved Oxygen Electrodes
Connect any ProDIGITAL integrated probe/cable assembly the ProSolo to form a complete sampling system. Probe assemblies are user-replaceable with cable lengths ranging from 1 to 100 meters.
  • Dissolved oxygen technology has advanced from electrochemical sensors, such as polarographic and galvanic to luminescent-based optical sensors
  • Digital smart sensors
  • Single cable design for probe assembly lengths ranging from 1 to 100 meters
The ODO®/CT probe assemblies pair with the ProSolo Handheld to form a complete sampling system. The ODO®/CT has all theMore Product Information