ProSolo Cable Options, ODO®/T Cable Assemblies, YSI
Supplier: YSI
627200-10 627200-20 627200-30 627200-50 627200-100 627200-1 627200-4
76308-034EA 1780.28 CAD
76308-034 76308-036 76308-038 76308-040 76308-042 76308-358 76308-360
ProSolo Cable Options, ODO®/T Cable Assemblies, YSI
Electrodes Dissolved Oxygen Electrodes
Connect any ProDIGITAL integrated probe/cable assembly the ProSolo to form a complete sampling system. Probe assemblies are user-replaceable with cable lengths ranging from 1 to 100 m.
  • Dissolved oxygen technology has advanced from electrochemical sensors, such as polarographic and galvanic to luminescent-based optical sensors
  • Digital smart sensors
  • Single cable design for probe assembly lengths ranging from 1 to 100 m
The ODO®/T probe assemblies pair with the ProSolo Handheld to form a complete sampling system. It features integral, digital sensors,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
DO concentration 0 - 50 mg/l
DO saturation 0 - 500%
DO accuracy 0 to 200%: 1% of reading or 1% saturation, whichever is greater; 200 to 500%: ±8% of reading; 0 to 20 mg/l: ±0.1 mg/l or 1% of reading, whichever is greater; 20 to 50 mg/l: ±8% of reading
DO resolution 0.01 mg/l and 0.1%, or 0.1 mg/l and 1% (auto-adjusts based on range)