5-Carboxytetramethylrhodamine (5-TAMRA) free acid amine-reactive fluorescent dye

Supplier: Biotium

90009
89139-532EA 324.96 CAD
89139-532
5-Carboxytetramethylrhodamine (5-TAMRA) free acid amine-reactive fluorescent dye
5-Carboxytetramethylrhodamine (5-TAMRA)

Free acid form of 5-TAMRA (5-Carboxytetramethylrhodamine, single isomer).


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO, DMF, or MeOH
  • Store at 4°C and protect from light, especially in solution

Formula: C₂₅H₂₂N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00269769
CAS Number: 91809-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR