Agarose IV, powder, Highest Gel Strength for biotechnology

Supplier: G-Biosciences

RC-011 RC-012
CA71003-336EA 357.85 CAD
CA71003-336 CA71003-338
Agarose IV, powder, Highest Gel Strength for biotechnology
Agarose

G-Biosciences offers several different biotechnology-grade agarose powders. Agarose IV is a linear polymer with a very high molecular weight, giving gel structures unlike those of traditional agaroses. This characteristic produces a strong intercatenary interaction, yeilding a gel with very high gel strength and higher exclusion limit.


For improved resolution, we recommend storing the gel at 4°C for 30 minutes before use.


Application:
Conventional Electrophoresis
Pulsed Field Gel Electrophoresis: Due to its higher exclusion limit, large molecules can be separated
Blotting
Agarose Bead Preparation
Cell and Enzyme Immobilization

Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Moisture ≤ 7%
Ash ≤ 0.25%
EEO (Electroendosmosis) ≤ 0.12
Sulfate ≤ 0.12%
Clarity (NTU) ≤ 4
Gel Strength 1% (g/cm²) ≥ 1800
Gel Strength 1.5% (g/cm²) ≥ 3200
Gelling Temperature 3% (°C) ≤ 36±1.5
Melting Temperature 3% (°C) ≤ 88±1.5
DNase/ RNase Activity None Detected
Gel Background Very Low
Appropriate Agarose Concentrations - 1X TAE BUFFER
1.8% 200-5,000bp
0,01 400-10,000bp
0.8% 1,000-15,000bp
0.5% 2,000-20,000bp
0.3% 3,000-40,000bp


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR