Agarose II, powder, melt_low Biotechnology

Supplier: G-Biosciences

RC-007 RC-008
CA95057-710EA 670.33 CAD
CA95057-710 CA95057-712
Agarose II, powder, melt_low Biotechnology
Agarose

G-Biosciences offers several different biotechnology-grade agarose powders. Agarose II has a low melting and gelling temperature compared to standard agaroses. The low melting temperature allows for the recovery of undamaged nucleic acids below the denaturation temperature. The low gelling temperature ensures that the agarose will be in a liquid state at a temperature range where in-gel manipulations can be performed without prior extraction from the agarose slice.


  • Lower gel strength compared to standard agarose
  • Higher clarity
  • Easy to handle


Applications:
Electrophoresis of DNA Fragments >1000bp
Tissue and Cell Culture
Viral Plaque Assays

Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Moisture ≤ 7%
Ash ≤ 0.4%
EEO (electroendosmosis) ≤ 0.12
Sulfate ≤ 0.12%
Clarity 1.5% (NTU) ≤ 4
Gel Strength 1.5% (g/cm2) ≥ 500
Gelling Temperature 1.5% (ºC) 24-28
Melting Temperature 1.5% (ºC) ≤ 65.5
DNase/ RNase Activity None Detected
DNA Resolution ≥1000bp Finely resolved
Gel background Very low
Appropriate Agarose Concentrations - 1X TAE BUFFER
0.75 % 500-12,000bp
1.00 % 300-8,000bp
1.25 % 200-4,000bp
1.50 % 150-3,000bp
1.75 % 100-2,000bp
2.0 % 50-1,000bp
Appropriate Agarose Concentrations - 1X TBE BUFFER


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR