Agarose I, powder general purpose reagent, for biotechnology low electroendosmosis (EEO)

Supplier: G-Biosciences

RC-122 RC-005 RC-006 RC-149
CA71003-518EA 73.87 CAD
CA71003-518 CA95057-706 CA95057-708 CA95057-714
Agarose I, powder general purpose reagent, for biotechnology low electroendosmosis (EEO)
Agarose

G-Biosciences offers several different biotechnology-grade agarose powders. G-Biosciences' Agarose I has low electroendosmosis (EEO) and high electrophoresis mobility. It is suitable for nucleic acid analytical and preparative electrophoresis, blotting, and protein electrophoresis (i.e. radial immunodiffusion).


  • Extraordinary mechanical resistance for more reliable and easier handling
  • Greater thermal stability due to high hysteresis (difference between gelling and melting temp)
  • High transparency and visibility
  • Easy preparation


Applications
Analytical nucleic acid electrophoresis
Preparative nucleic acid electrophoresis
Blotting
Protein electrophoresis such as radial immunodiffusion

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Moisture ≤ 7%
Ash 0,04
EEO 0.05-0.13
Sulfate ≤ 0.1%
Clarity 1.5% (NTU) ≤ 3
Gel Strength 1% (g/cm2) ≥ 1200
Gel Strength 1.5% (g/cm2) ≥ 2500
Gelling Temperature 1.5% (ºC) 36 ± 1.5
Melting Temperature 1.5% (ºC) 88 ± 1.5
DNase/ RNase Activity None Detected
DNA Resolution ≥1000bp Finely resolved
Gel background Very low
Appropriate Agarose Concentrations - 1X TAE BUFFER
0.6 % 1,000-15,000bp
0.8 % 500-10,000bp
1.0 % 250-7,000bp
1.2 % 200-5,000bp
1.5 % 100-3,000bp
2.0 % 50-2,000bp
Appropriate Agarose Concentrations - 1X TBE BUFFER

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR