Replacement Shield Plus Visor, Portwest
Supplier: Portwest
PW99CLR
76105-174EA 14.38 CAD
76105-174
Replacement Shield Plus Visor, Portwest
Face Shields
Replacement visor for PW96. It also fits PW58 and PS58. Made from durable Polycarbonate with 1mm thickness.
  • Clear visor aluminium bound
  • For medium-energy impact protection
  • Protects from impacts, metal splashes and hot solid projections
  • Protection against Molten Metal
Replacement visor to be used on PW96 browguard support. Compatible with PW58 helmet visor carrier and PS58 helmet visor holder.