Wireless Tube Rack with RFID for CapTrack™ Systems, Globe Scientific
Supplier: Globe Scientific
456445R-R 456445W-R 456445Y-R 456440B-R 456440G-R 456440M-R 456440N-R 456440R-R 456440W-R 456440Y-R 456445B-R 456445G-R 456445M-R 456445N-R
76308-962EA 53.21 CAD
76308-962 76308-964 76308-966 76309-012 76309-014 76309-016 76309-018 76309-020 76309-022 76309-024 76309-026 76309-028 76309-030 76309-032
Wireless Tube Rack with RFID for CapTrack™ Systems, Globe Scientific
Racks Tube Racks
These RFID enabled racks specific for Captrack™ systems come pre-assembled with a unique RFID disk.
  • Fully autoclavable
  • Molded from reinforced nylon for strength and durability with no coatings to chip, peel, or rust
  • Single piece, three (3) tier grid design for secure and stable racking of tubes
  • Good temperature and chemical resistance to withstand a variety of lab procedures
  • Large, flat endplates for labeling and easy sample identification
After processing samples with the CapTrack™ benchtop recapping archiver, these racks allow users to easily trackMore Product Information