CapTrack™ Benchtop Recapping Archiver, Globe Scientific
Supplier: Globe Scientific
CT1 CT2 CR1
76308-954EA 178495.62 CAD
76308-954 76308-956 76308-958
CapTrack™ Benchtop Recapping Archiver, Globe Scientific
Crimpers, Cappers and Decappers
CapTrack™ is the ideal benchtop solution for increased efficiency, sample integrity and safety in blood processing labs.
  • CT1: Automated tube recapping, tracking and sample management with tube height and cap color recognition recall function
  • CT2: Automated tube recapping, tracking and sample management with rack position recall function
  • CR1: Automated tube recapping
CapTrack™ automation eliminates the need for time-consuming manual processing that can result in reduced efficiency and increased risk of sample contamination. It gives technicians theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable