VITsafe™ Safety Wash Bottles, Brandtech
Supplier: Brandtech
1352899 1352909 1352959 1352819 1352869 1452829
76307-364PK 252.59 CAD
76307-364 76307-366 76307-368 76307-444 76307-446 76307-448
VITsafe™ Safety Wash Bottles, Brandtech
Bottles Wash Bottles
Wide Mouth VITsafe™ safety wash bottles are made from low-density polyethylene (or Polypropylene for the Acetone bottle). Wash tubes are made of polypropylene.
  • Durable safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Name in German, English, French, and Spanish
  • Printed with chemical formula, CAS No., hazard pictogram and signal word
  • Printed with risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Metal-free, color coded VENT-Caps reduce static overpressure, virtually eliminating leaks or drips from the dispensing tube. Wide-mouth openings simplify filling, even without a funnel.