VWR® Sterile T-Shaped Cell Spreaders
Supplier: VWR International
76299-566 76299-568 76299-570
76299-566CS 1263.83 CAD
76299-566 76299-568 76299-570
VWR® Sterile T-Shaped Cell Spreaders
Spreaders Cell Spreaders
VWR® disposable T-shaped Cell Spreaders are ideal for spreading bacterial cultures evenly.
  • Spread and disperse liquids across the surface of culture plates
  • Smooth edges minimize 'gouging'
  • Sterilized by EO irradiation
  • BSE/TSE-free, and latex-free
VWR® T-Shaped Cell Spreaders are manufactured under EN ISO 13485:2016 and ISO 9001:2015 quality management systems.
Certifications: ISO 13485 :2003 and ISO 9001: 2008.