Magnesium chloride hexahydrate FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesium dichloride

Management of Change (MOC) category = RL

MACR7550-32 MACR7550-32
CAMK7550-32EA 371.43 CAD
CAMK7550-32 MK7550-32
Magnesium chloride hexahydrate FCC, Macron Fine Chemicals™
Magnesium chloride hexahydrate

(Abbott code 1418)

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Appearance Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Assay (MgCl₂.6H₂O) 99.0 - 105.0 %
FCC - Ammonium (NH₄) ≤0.005 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Sulfate (SO₄) ≤0.03 %
Solubility Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR