OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler Toledo
11123584 11123585 11123586 11123587 11123589 11123590 11123591 11123592 11123607 11123608 11123609 11123610 11123611 11123612 11123613 11123614 11123615 11123621 11123593 11123596 11123598 11123601 11123603 11123604 11123618 11123619 11123620 11123622 11123623 11123624 11123626 11123594 11123597 11123599 11123602 11123617 11123595 11123600 11123616 11123627 11123625
CA97007-486EA 102.97 CAD
CA97007-486 CA97007-488 CA97007-490 CA97007-492 CA97007-494 CA97007-496 CA97007-498 CA97007-500 CA97007-502 CA97007-528 CA97007-530 CA97007-532 CA97007-534 CA97007-536 CA97007-538 CA97007-540 CA97007-542 CA97007-544 CA97007-556 97007-504 97007-510 97007-514 97007-520 97007-524 97007-526 97007-550 97007-552 97007-554 97007-558 97007-560 97007-562 97007-566 97007-506 97007-512 97007-516 97007-522 97007-548 97007-508 97007-518 CA97007-546 CA97007-568 CA97007-564
OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.