Eosin Y (yellowish) 1% (w/v) alcoholic solution for histology
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Tetrabromofluorescein disodium salt
, Eosin Y disodium salt
, Eosin 225
, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt
, Solvent Red 43
, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate
, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Field's Stain B
, 2,4,5,7-Tetrabromofluorescein disodium salt
R2850000-1A
R2850000-4A
R2850000-500A
RC285032EA
285.32
CAD
RC285032
RC2850-1
RC285016
Eosin Y (yellowish) 1% (w/v) alcoholic solution for histology
Eosin Y (yellowish)
Formula:
C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 UN: 1987 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...