J.T.Baker® BAKERBOND®, Poly Hi-Propyl Mixed Mode Hydrophobic Interaction Chromatography Resin
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
The J.T.Baker® BAKERBOND® Poly Hi-Propyl mixed mode hydrophobic interaction chromatography (HIC) resin provides improved selectivity for superior separation in process chromatography applications.
- Store between 2 and 15 °C
Powered by a proprietary surface chemistry combining hydrophobic functionality with weak anionic exchange sites, BAKERBOND® Poly Hi-Propyl mixed mode hydrophobic interaction chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® Poly Hi-Propyl ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Ordering information: 1 mL and 5 mL lab columns are also available under complementary products.