Minum-Ware® Bulb Immersion Thermometers, Non-Mercury, Chemglass
Supplier: Chemglass
MW-110-N-01 MW-110-N-02
66025-544EA 133.5 CAD
66025-544 66025-546
Minum-Ware® Bulb Immersion Thermometers, Non-Mercury, Chemglass
Thermometers Analog Thermometers
Bulb immersion, mineral spirit filled, thermometer with engraved stem.
The stem O.D is 5.5 to 6.0 mm.
Delivery information: Each thermometer is supplied with a #106 Viton O-ring for suspension of thermometer in open distillation systems.