Formaldehyde 37% in aqueous solution stabilized ACS
Supplier: RICHARD-ALLAN SCIENTIFIC CA
|
Danger
|
Stabilization:
Stabilised with methanol
9300-5
9300-1
CA48218-722EA
528.91
CAD
CA48218-722
CA48218-724
Formaldehyde 37% in aqueous solution stabilized ACS
Formaldehyde
Not intended for use as a disinfectant.
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 2209 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...