Antimony, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: Avantor
VWR Chemicals BDH® ARISTAR®
CABDH89800-268 BDH82026-222 BDH89800-016 CABDH89800-014 BDH89800-270 CABDH89800-270 BDH89800-268 BDH82026-050 BDH82026-052 CABDH82026-050 CABDH82026-052 BDH89800-014 BDH82026-220 CABDH82026-222 CABDH82026-220
CABDH89800-268EA 172.45 CAD
BDH82026-050 BDH89800-270 BDH82026-220 BDH82026-052 CABDH89800-268 BDH82026-222 CABDH89800-014 CABDH82026-052 CABDH82026-050 CABDH89800-270 CABDH82026-222 BDH89800-268 BDH89800-016 CABDH82026-220 BDH89800-014
Antimony, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Standards Cationic Standards Antimony Standards
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Long shelf life
  • NIST traceable
  • Precise and accurate
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS). Specific lot C of A’s can also be found online.
Enhanced packaging extends the shelf life to 4 years by sealing the product in an external aluminized bag over the regular poly bottle.
This specially designed outer bag prevents transpiration (the loss of water, or solvent, through the container.) Therefore, most of these standards have a 4 year shelf life until the outer bag is opened. After opening the outer bag, the customer will mark the inner bottle with a one-year shelf life from that date.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 µg/mL
Please note: 1000 ppm = 1000 µg/mL