Rhodium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: Avantor
VWR Chemicals BDH® ARISTAR®
CABDH89800-442 BDH89800-228 BDH89800-442 CABDH89800-256 BDH89800-256 CABDH89800-130 BDH89800-126 CABDH82026-218 CABDH89800-228 BDH89800-440 BDH82026-218 BDH82026-044 CABDH82026-216 CABDH82026-044 CABDH89800-440 CABDH89800-128 BDH89800-128 BDH82026-216 CABDH89800-126 BDH89800-130
CABDH89800-228EA 313.33 CAD
CABDH89800-228 BDH82026-044 CABDH89800-128 CABDH89800-126 CABDH89800-442 CABDH89800-256 CABDH89800-440 BDH82026-216 CABDH89800-130 BDH89800-228 BDH82026-218 BDH89800-128 BDH89800-126 CABDH82026-044 BDH89800-442 BDH89800-256 BDH89800-440 CABDH82026-218 BDH89800-130 CABDH82026-216
Rhodium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Standards Cationic Standards Rhodium Standards
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Long shelf life
  • NIST traceable
  • Precise and accurate
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS). Specific lot C of A’s can also be found online.
Enhanced packaging extends the shelf life to 4 years by sealing the product in an external aluminized bag over the regular poly bottle.
This specially designed outer bag prevents transpiration (the loss of water, or solvent, through the container.) Therefore, most of these standards have a 4 year shelf life until the outer bag is opened. After opening the outer bag, the customer will mark the inner bottle with a one-year shelf life from that date.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 µg/mL
Please note: 1000 ppm = 1000 µg/mL