Dura-Mag™ Magnetic Stirrers, 5-position, Chemglass
Supplier: Chemglass
CLS-4100-04 CLS-4100-06 CLS-4100-A4 CLS-4100-A6
30624-010EA 7701.39 CAD
30624-010 30624-012 30624-018 30624-020
Dura-Mag™ Magnetic Stirrers, 5-position, Chemglass
Stirrers Magnetic Hotplate Stirrers, Multi-position
The five position magnetic stirrer is a rugged design for 24/7 laboratory or production applications. It is ideal for CO2 incubators with a maximum capacity of five 1000 ml spinner vessels.
  • Trouble free brushless motor
  • Heavy duty magnets and drive system
  • Stainless steel top with easy alignment marks
  • Non-slip trouble free toothed belt design for long life
  • Illuminated LCD display
The factory installed alarm has 85 dB internal audible buzzer, serial port for external alarm configuration (external alarm is not supplied), convenient alarm reset button andMore Product Information