Accucore™ Biphenyl Reversed Phase LC Columns, Thermo Scientific
Supplier: Thermo Scientific
17826-012105 17826-052130 17826-102130
76292-194PK 887.45 CAD
76292-194 76292-196 76292-198
Accucore™ Biphenyl Reversed Phase LC Columns, Thermo Scientific
Chromatography Columns
Thermo Scientific™ Accucore™ Biphenyl LC columns are designed for fast and reliable separation of critical pairs and isomers. These columns provide unique selectivity for aromatic and moderately polar analytes. Their optimized packing offers a rugged platform for a variety of matrices.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Mixed-mode selectivity for aromatic and moderately polar analytes
  • Enhanced Pi-pi interactions with aromatics
  • Excellent lot-to-lot reproducibility
Based on Core Enhanced Technology, Accucore™ liquid chromatography reversed phase columns provide fast, high-resolution separations without the elevated backpressures seen using sub-2 µm particles.
These columns offer fast run times, excellent robustness, and consistent reproducibility, and provide the perfect balance of polar analyte retention and hydrophobic analyte elution.