Dewar Flasks with Handles, Pope Scientific
Supplier: Pope Scientific
8921 8942 8945
63422-110EA 1627.94 CAD
63422-110 63422-142 63422-063
Dewar Flasks with Handles, Pope Scientific
Dewars and Cryogenic Vessels
These flasks are ideal for temporary storage and liquid gas service, especially liquid nitrogen. The cylindrical, wide mouth design with the fully silvered borosilicate glass inner flask incorporates the convenience of a coated handle.
  • Aluminum metal housing extends full length of flask for added protection
  • Vacuumed to 5 torr for efficiency
  • Extended lip to facilitate pouring
  • Bottom of base has a cushioned rim for added stability
  • Polyethylene stopper, specially vented, minimizes evaporation
All models use the fitted caps for extended holding.