Accessories for Boekel Tube Rotator
Supplier: Boekel
260652 260653 260655 260654 260651
97043-542EA 355.45 CAD
97043-542 97043-544 97043-546 97043-548 97043-540
Accessories for Boekel Tube Rotator
Shakers Rotating/Rocking Shakers
Order Now
SPECIFICATIONS
Environmentally Preferable
Capacity 46 9-11 mm tubes, 28 14-18 mm tubes, 12 27-34 mm tubes, 65 18.5 mm culture tubes (drum rotor) and 65 14 mm culture tubes (drum rotor)
Speed 4-80 rpm
Angle 90 - 180°
Dimensions 304 x 431 x 381 mm (12 x 17 x 15")
Electrical 19.4 vdc, 4.74A via 100-240 AC adapter drum
Timer Up to 99 hrs. and 59 min.