ZymoTaq DNA Polymerase, Zymo Research
Supplier: Zymo Research
E2001
E2002
77001-902EA
135.93
CAD
77001-902
76211-540
ZymoTaq DNA Polymerase, Zymo Research
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
ZymoTaq DNA polymerase is a heat-activated, 'hot start' polymerase that has 3′-terminal transferase activity. The addition of 'A' overhangs to amplified DNA makes it ideal for use in TA-cloning.
- Simple and easy to use
- Compatible with real-time, quantitative PCR, and suitable for TA-cloning
- Generates specific amplicons with little or no by-product formation
Reduces non-specific PCR product formation from difficult templates such as bisulfite converted DNA.