Gyrolab™ Wash Buffer and Rexxip Buffers, Gyros Protein Technologies
Supplier: GYROS US INC. MS
P0004823 P0004825 P0004821 P0020027 P0004994
MSPP-P0004823EA 1332.31 CAD
MSPP-P0004823 MSPP-P0004825 MSPP-P0004821 MSPP-P0020027 MSPP-P0004994
Gyrolab™ Wash Buffer and Rexxip Buffers, Gyros Protein Technologies
Buffers Electrophoresis Buffers Washing Buffers
Gyrolab™ wash buffer (pH 11) is a biodegradable high pH washing buffer that is highly efficient when used in combination with Rexxip buffers.
  • Improve data quality
  • Cut assay development time
  • Compatible with a variety of molecules with different analytical properties
An additional needle wash with Gyrolab™ wash buffer after sample and reagent transfer can greatly improve the quality of data, especially for analytes that are positively-charged.
To prepare the buffer, dissolve 10 g of Gyrolab™ wash buffer in one liter of deionized or distilled water and filter the solution through a 0.22 μm filter.